Learn More
Charter Medical FREEZE PAK, 250ML MAX FILL V
Catalog No. NC2144127
Encompass
250mL Max Fill Volume, (2) FP-FLEX Lines (8in L, 1/8in ID x 1/4in OD) Sealed 25EA/CS
This item is not returnable. View return policy
Catalog No. NC2144127 Supplier Charter Medical Supplier No. FPFLEX250A
250mL Max Fill Volume, (2) FP-FLEX Lines (8in L, 1/8in ID x 1/4in OD) Sealed 25EA/CS