Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96 Deep-Well Microtiter Plates or 1 3/4" Max H Boxes, 6 x 6 Array
Catalog No. 501960682 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely keep plates and boxes in place. Each compartment holds a single plate or box. Plates and boxes can be accessed from either side. The racks accommodate freezers with left- or right-handed doors.
Catalog No. 50-196-0682 Supplier Cole-Parmer Supplier No. 0439836
These racks come with locking rods to securely keep plates and boxes in place. Each compartment holds a single plate or box. Plates and boxes can be accessed from either side. The racks accommodate freezers with left- or right-handed doors.