Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Standard 2", 3" or 3.75" Boxes, 4 columns; 9 3/8"H
Catalog No. 501960493 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks are great for storing standard 2-, 3- and 3.75 inch cardboard, plastic or metal boxes. Designed to fit all major brands of f 80C ultralow upright freezers. The racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0493 Supplier Cole-Parmer Supplier No. 0439833
These upright freezer racks are great for storing standard 2-, 3- and 3.75 inch cardboard, plastic or metal boxes. Designed to fit all major brands of f 80C ultralow upright freezers. The racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.