Learn More
Electron Microscopy Sciences EMS Freeze Substitution Kit with Shaker
SDP
Catalog No. 501902378
Encompass_Preferred
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete. With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
Catalog No. 50-190-2378 Supplier Electron Microscopy Sciences Supplier No. 34500
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete. With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.