Learn More
Caron Products And Services Freeze/Thaw Chamber, 33 cu. ft., Temp & RH
Catalog No. NC2676690
Encompass_Preferred
33 ft3 (934 L) Freeze/Thaw Chamber offers programmable temperature (-25C to 70C) and humidity settings (20-98% RH). Cycles between setpoints, enabling continuous simulation of freezing and thawing for rapid testing. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.
Catalog No. NC2676690 Supplier Caron Products And Services Supplier No. 7900332
Add to Cart
Add to Cart
Request a Quote
33 ft3 (934 L) Freeze/Thaw Chamber offers programmable temperature (-25C to 70C) and humidity settings (20-98% RH). Cycles between setpoints, enabling continuous simulation of freezing and thawing for rapid testing. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.