Learn More
Electron Microscopy Sciences 144 Place Freezer Storage Box
SDP
Catalog No. 501928516
Encompass_Preferred
144-place PCR Freezer Box fits in a standard 2 inch freezer rack. Designed for ULT freezer storage at temperatures down to -86 degree C, it is durable and impervious to moisture. The clear polycarbonate lid and base and alpha-numeric index make it easy to identify samples within the box. Two boxes may be stacked to fit together in one space in a 2 inch freezer storage rack if using flat-capped PCR tubes.
Catalog No. 50-192-8516 Supplier Electron Microscopy Sciences Supplier No. 61605
144-place PCR Freezer Box fits in a standard 2 inch freezer rack. Designed for ULT freezer storage at temperatures down to -86 degree C, it is durable and impervious to moisture. The clear polycarbonate lid and base and alpha-numeric index make it easy to identify samples within the box. Two boxes may be stacked to fit together in one space in a 2 inch freezer storage rack if using flat-capped PCR tubes.