Learn More
Thomas Scientific 2" White Freezer Box w/ 81 Place Divider, No Drain Holes, 5 1/4" x 5 1/4" x 2"
SDP
Catalog No. NC9864798
Encompass_Preferred
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2
Catalog No. NC9864798 Supplier Thomas Scientific Supplier No. 1149H28
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2