Learn More
TWD Scientific LLC Freezer Box, Twdbiox, 2In, 100 Well, Polypropylene, Assorted Colors, Removable Lid, Numeric Grid On Lid And Box, 20/Case.
SDP
Catalog No. 502285099 Shop All TWD Scientific LLC Products
Encompass_Preferred
2 Inch 100 Well Polypropylene Freezer Box (Dimensions: 132X132X53Mm) With Removable Lid. Boxes/Lids Have Numeric Grid For Identification Purposes. Boxes Come In Assorted Colors (Clear, Pink, Blue, Green And Yellow). 20/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5099 Supplier TWD Scientific LLC Supplier No. BX909200
2 Inch 100 Well Polypropylene Freezer Box (Dimensions: 132X132X53Mm) With Removable Lid. Boxes/Lids Have Numeric Grid For Identification Purposes. Boxes Come In Assorted Colors (Clear, Pink, Blue, Green And Yellow). 20/Case, Price/Case