Learn More
TWD Scientific LLC Freezer Box, Twdbiox, 2In, 25 Well, Polypropylene, Assorted Colors, Removable Lid, Numeric Grid On Lid And Box, 20/Case.
SDP
Catalog No. 502285097 Shop All TWD Scientific LLC Products
Encompass_Preferred
2 Inch 25 Well Polypropylene Freezer Box (Dimensions: 75X75X52Mm) With Removable Lid. Boxes/Lids Have Numeric Grid For Identification Purposes. Boxes Come In Assorted Colors (Clear, Pink, Blue, Green And Yellow). 30/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5097 Supplier TWD Scientific LLC Supplier No. BX9090251
2 Inch 25 Well Polypropylene Freezer Box (Dimensions: 75X75X52Mm) With Removable Lid. Boxes/Lids Have Numeric Grid For Identification Purposes. Boxes Come In Assorted Colors (Clear, Pink, Blue, Green And Yellow). 30/Case, Price/Case