Learn More
JDM LAB SOLUTIONS LLC Freezer box for 5mL MacroTube, 25 place
Catalog No. NC4039640
Encompass
Cardboard freezer box with hinged lid, 3 inch, with insert for 25 snap-cap 5mL MacroTubes, 5.25 x 5.25 x 3 inches, 5/pk
Catalog No. NC4039640 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMC258025
Cardboard freezer box with hinged lid, 3 inch, with insert for 25 snap-cap 5mL MacroTubes, 5.25 x 5.25 x 3 inches, 5/pk