Learn More
Cole-Parmer Argos Technologies PolarSafe 81-Place Freezer Boxes, Orange, PP; 50/CS
Catalog No. 501960620 Shop All Cole-Parmer Products
Encompass_Preferred
Boxes withstand temperatures from -80C to 121C, and will not degrade with repeated thaws. Each box features an alphanumeric grid on the exterior and interior, stacking rings, and a matte surface suitable for markers or labels. Box top is clear for easy visual inspection of samples and bottoms are colored for additional identification and organization. Boxes accommodate 0.5, 1.5, and 2.0 mL microtubes, and most brands of cryogenic vials.
Catalog No. 50-196-0620 Supplier Cole-Parmer Supplier No. 2445285
Boxes withstand temperatures from -80C to 121C, and will not degrade with repeated thaws. Each box features an alphanumeric grid on the exterior and interior, stacking rings, and a matte surface suitable for markers or labels. Box top is clear for easy visual inspection of samples and bottoms are colored for additional identification and organization. Boxes accommodate 0.5, 1.5, and 2.0 mL microtubes, and most brands of cryogenic vials.