Learn More
Cole-Parmer Argos Technologies PolarSafe Metal Freezer Box, Aluminum, 2"; 1/Ea
Catalog No. 501960680 Shop All Cole-Parmer Products
Encompass_Preferred
These metal freezer boxes are guaranteed not to rust. They feature drain holes that is ideal for storing samples in most liquid nitrogen tanks offered on the market. They come in stainless steel or aluminum, and the boxes can be stored in the same rack configurations as the two and three inch cardboard boxes. Cardboard dividers can be added, too, depending on the application.
Catalog No. 50-196-0680 Supplier Cole-Parmer Supplier No. 0439810
These metal freezer boxes are guaranteed not to rust. They feature drain holes that is ideal for storing samples in most liquid nitrogen tanks offered on the market. They come in stainless steel or aluminum, and the boxes can be stored in the same rack configurations as the two and three inch cardboard boxes. Cardboard dividers can be added, too, depending on the application.