Learn More
MTC Bio Cardboard freezer box with hinged lid, 3 inch, with insert for 36 screw-cap 5mL MacroTubes, 5.25 x 5.25 x 3 inches, 5/pk
Catalog No. NC1905726
Encompass_Preferred
MTC Bio’s 5mL MacroTube Cardboard Freezer Boxes are made of recyclable material, with a liquid-resistant outer coating. They are shipped fully assembled and are available in a 5×5 version that holds 25 x 5mL Snap-Cap tubes or a 6×6 version that holds 36 X 5mL Screw-Cap tubes. Each box has liquid nitrogen drainage slots and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.). Attached hinged lid. 5×5 version for 5mL Snap-Caps. 6×6 version for 5mL Screw-Caps. Alpha-numeric coding. Polycarbonate box fits 25 x 5mL tubes. Polycarbonate box usable temp range is -196° to +121°
Catalog No. NC1905726 Supplier MTC Bio Supplier No. C258036
MTC Bio’s 5mL MacroTube Cardboard Freezer Boxes are made of recyclable material, with a liquid-resistant outer coating. They are shipped fully assembled and are available in a 5×5 version that holds 25 x 5mL Snap-Cap tubes or a 6×6 version that holds 36 X 5mL Screw-Cap tubes. Each box has liquid nitrogen drainage slots and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.). Attached hinged lid. 5×5 version for 5mL Snap-Caps. 6×6 version for 5mL Screw-Caps. Alpha-numeric coding. Polycarbonate box fits 25 x 5mL tubes. Polycarbonate box usable temp range is -196° to +121°