Learn More
Chemglass Life Sciences Freezer Box, Hinged Lid, Green, 100-Place, Complete with Dividers
SDP
Catalog No. 501979420
Encompass_Preferred
Freezer Box, Hinged Lid, Green, 100-Place, Complete with Dividers. New FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81 place or 100 place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.). 5-1/4 x 5-1/4 x 2 Inches - Standard Size, for Existing Racks. Alpha-Numeric Coding - Top, Sides and Bottom
Catalog No. 50-197-9420 Supplier Chemglass Life Sciences Supplier No. CLS1197100G
Freezer Box, Hinged Lid, Green, 100-Place, Complete with Dividers. New FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81 place or 100 place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.). 5-1/4 x 5-1/4 x 2 Inches - Standard Size, for Existing Racks. Alpha-Numeric Coding - Top, Sides and Bottom