Learn More
Electron Microscopy Sciences KeepIT Freezer Box 25, White, 75 x 75 x 52 mm
SDP
Catalog No. 501929338
Encompass_Preferred
KeepIt Freezer Boxes provide an ideal method for batching and storing samples. They match the colors of CryoELITE Cryogenic Vials. Made from Eastman Tritan BPA free, shatter resistant resin. Feature openings in bottom to facilitate scanning CryoELITE 2D data matrix bar code inserts. Standard footprint compatible with liquid nitrogen storage shelves and freezer drawers. Accommodates up to 25 internal or external threaded cryogenic vial sizes 1.2 - 2ml.
Color: White
Dimensions: 75 x 75 x 52 mm
Catalog No. 50-192-9338 Supplier Electron Microscopy Sciences Supplier No. 6160625W
KeepIt Freezer Boxes provide an ideal method for batching and storing samples. They match the colors of CryoELITE Cryogenic Vials. Made from Eastman Tritan BPA free, shatter resistant resin. Feature openings in bottom to facilitate scanning CryoELITE 2D data matrix bar code inserts. Standard footprint compatible with liquid nitrogen storage shelves and freezer drawers. Accommodates up to 25 internal or external threaded cryogenic vial sizes 1.2 - 2ml.
Color: White
Dimensions: 75 x 75 x 52 mm