Learn More
Cole-Parmer Argos Technologies PolarSafe Freezer Box, PC, 81-Place, 2", 12.5 mm; 4/Pk
Catalog No. 501960605 Shop All Cole-Parmer Products
Encompass_Preferred
These polycarbonate freezer boxes are available with 25, 81 and 100-place divisions. They feature bottom drain holes for safe venting. The 25- and 81-place boxes have individual cell compartment dividers, while the 100-place box holds tubes in a pegged grid divider.
Catalog No. 50-196-0605 Supplier Cole-Parmer Supplier No. 0439805
These polycarbonate freezer boxes are available with 25, 81 and 100-place divisions. They feature bottom drain holes for safe venting. The 25- and 81-place boxes have individual cell compartment dividers, while the 100-place box holds tubes in a pegged grid divider.