Learn More
Cole-Parmer Argos Technologies PolarSafe Freezer Box, PC, 81-Place, 3", 12.5 mm; 4/Pk
Catalog No. 501960678 Shop All Cole-Parmer Products
Encompass_Preferred
Polycarbonate freezer boxes, available with 25, 81, and 100-place divisions, feature bottom drain holes for safe venting. The 25- and 81-place boxes have individual cell compartment dividers, while the 100-place box holds tubes in a pegged grid divider. Designed to withstand temperatures from -320.8 to 250F or -196 to 121C.
Catalog No. 50-196-0678 Supplier Cole-Parmer Supplier No. 0439806
Polycarbonate freezer boxes, available with 25, 81, and 100-place divisions, feature bottom drain holes for safe venting. The 25- and 81-place boxes have individual cell compartment dividers, while the 100-place box holds tubes in a pegged grid divider. Designed to withstand temperatures from -320.8 to 250F or -196 to 121C.