Learn More
Cole-Parmer Argos Technologies PolarSafe Metal Freezer Box, Stainless Steel, 2"; 1/Ea
Catalog No. 501960606 Shop All Cole-Parmer Products
Encompass_Preferred
These metal freezer boxes are guaranteed not to rust. They feature drain holes, making them ideal for storing samples in most liquid nitrogen tanks offered on the market. They are available in stainless steel or aluminum and can be stored in the same rack configurations as the 2 and 3 cardboard boxes. Cardboard dividers can be added depending on the application.
Catalog No. 50-196-0606 Supplier Cole-Parmer Supplier No. 0439808
These metal freezer boxes are guaranteed not to rust. They feature drain holes, making them ideal for storing samples in most liquid nitrogen tanks offered on the market. They are available in stainless steel or aluminum and can be stored in the same rack configurations as the 2 and 3 cardboard boxes. Cardboard dividers can be added depending on the application.