Learn More
USA Scientific Inc 100-place cardboard box for 12 x 75 mm tubes
Catalog No. NC9624177
Encompass
100-place cardboard box for 12 x 75 mm tubes, deep lid, 5 1/4 inch L x 5 1/4 inch W x 2 15/16 inch H, includes drainch holes inch base
Catalog No. NC9624177 Supplier USA Scientific Inc Supplier No. 90233100
100-place cardboard box for 12 x 75 mm tubes, deep lid, 5 1/4 inch L x 5 1/4 inch W x 2 15/16 inch H, includes drainch holes inch base