Learn More
Research Products International Corp Freezer Drawer Rack for 15ml Tubes, 120x15ml
SDP
Catalog No. 501977414
Encompass
Stainless steel freezer racks feature easy slide out drawers to provide quick and convenient access to samples. Racks accommodate a large number of tubes for improved freezer capacity and eliminate the need for storing individual boxes. Racks fit standard upright freezers.
Catalog No. 50-197-7414 Supplier Research Products International Corp Supplier No. 181505
Stainless steel freezer racks feature easy slide out drawers to provide quick and convenient access to samples. Racks accommodate a large number of tubes for improved freezer capacity and eliminate the need for storing individual boxes. Racks fit standard upright freezers.