Learn More
Research Products International Corp FREEZER DRAWER RACK, 15 & 50ML
SDP
Catalog No. 501367686
Encompass
Stainless steel freezer racks feature easy slide out drawers to provide quick and convenient access to samples. Racks accommodate a large number of tubes for improved freezer capacity and eliminate the need for storing individual boxes. Drawers can be interchanged within the same rack for combination storage of 60 15ml and 30 50ml tubes.
Racks fit standard upright freezers.
Catalog No. 50-136-7686 Supplier Research Products International Corp Supplier No. 181525
Stainless steel freezer racks feature easy slide out drawers to provide quick and convenient access to samples. Racks accommodate a large number of tubes for improved freezer capacity and eliminate the need for storing individual boxes. Drawers can be interchanged within the same rack for combination storage of 60 15ml and 30 50ml tubes.
Racks fit standard upright freezers.