Learn More
Alkali Scientific Alkali Scientific™ Freezer Drawer Racks for Storage of Bottles, 1 Drawer, Drawer Area Capacity: 20 5/8″ x 4 1/2″
SDP
Catalog No. 503028030
Encompass_Preferred
For Storage Bottles The UFDR-LB10-1 Upright Freezer Drawer Rack for storage bottles features 1 drawer and is constructed of corrosion resistant stainless steel. Drawer racks facilitate both access to and storage of bottles without having to remove the whole rack from the freezer. These freezer racks for storage bottles include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Dimensions L x H x W : 22 3/16 x 4 1/2 x 5 1/2 inch (564 x 114 x 140mm)Capacity: Drawer Area 20 5/8 x 4 1/2 (Per Drawer) 523 x 114mm
Catalog No. 50-302-8030 Supplier Alkali Scientific Supplier No. UFDRLB101
For Storage Bottles The UFDR-LB10-1 Upright Freezer Drawer Rack for storage bottles features 1 drawer and is constructed of corrosion resistant stainless steel. Drawer racks facilitate both access to and storage of bottles without having to remove the whole rack from the freezer. These freezer racks for storage bottles include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Dimensions L x H x W : 22 3/16 x 4 1/2 x 5 1/2 inch (564 x 114 x 140mm)Capacity: Drawer Area 20 5/8 x 4 1/2 (Per Drawer) 523 x 114mm