Learn More
MSC Zep Cooler and Freezer Cleaner, Low Freezing, 5 gal.
Catalog No. 502818181
Encompass
This specially formulated, ready-to-use liquid is designed for cleaning in sub-freezing temperatures, eliminating the need for defrosting. It maintains its liquid state down to -40°F and can be diluted for economy at -30°F (1:1) and +10°F (1:2). This non-flammable cleaner is authorized by the USDA for use on non-food contact surfaces in federally inspected meat and poultry processing plants.
  • Ready-to-use formulation, eliminating mixing; dilutable for greater economy.
  • Remains liquid at temperatures as low as -40°F.
  • Non-flammable, eliminates a potential hazard.
  • No odor, will not impart flavor or odor to food products.
  • Requires no rinse when used as directed.
  • Safe on all metals.
  • USDA authorized for use in federally inspected meat and poultry processing plants.
Catalog No. 50-281-8181 Supplier MSC Supplier No. 17640707
This specially formulated, ready-to-use liquid is designed for cleaning in sub-freezing temperatures, eliminating the need for defrosting. It maintains its liquid state down to -40°F and can be diluted for economy at -30°F (1:1) and +10°F (1:2). This non-flammable cleaner is authorized by the USDA for use on non-food contact surfaces in federally inspected meat and poultry processing plants.
  • Ready-to-use formulation, eliminating mixing; dilutable for greater economy.
  • Remains liquid at temperatures as low as -40°F.
  • Non-flammable, eliminates a potential hazard.
  • No odor, will not impart flavor or odor to food products.
  • Requires no rinse when used as directed.
  • Safe on all metals.
  • USDA authorized for use in federally inspected meat and poultry processing plants.