Learn More
Darwin Chambers Company Single Door Freeze-Thaw Chamber 21 cu.ft.
SDP
Supplier:  Darwin Chambers Company TH030SS
Encompass_Preferred
The TH030-SS is a 23.9 cubic feet single door freeze thaw chamber. Temperature range is -25°C to 50°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Freeze-thaw chambers are excellent for customers requiring ramp/soak programming. They exceed ICH/FDA guidelines concerning uniformity requirements. Approximate ramping ability is as follows 1°C per 2 minutes from -23°C to +23°C and back, unloaded. Note: Some defrost tolerance may be required for extended operation (more than 48 hours) below 4°C.
Catalog No. 50-189-3878
Add to Cart
Request a Quote