Learn More
Darwin Chambers Company Triple Door Freeze-Thaw Chamber 66 cu. ft.
SDP
Catalog No. 501893880
Encompass_Preferred
The TH084-SS is a 76.2 cubic feet triple door freeze thaw chamber. Temperature range is -25°C to 50°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Freeze-thaw chambers are excellent for customers requiring ramp/soak programming. They exceed ICH/FDA guidelines concerning uniformity requirements. Approximate ramping ability is as follows 1°C per 2 minutes from -23°C to +23°C and back, unloaded. Note: Some defrost tolerance may be required for extended operation (more than 48 hours) below 4°C.
Catalog No. 50-189-3880 Supplier Darwin Chambers Company Supplier No. TH084SS
Add to Cart
Add to Cart
Request a Quote
The TH084-SS is a 76.2 cubic feet triple door freeze thaw chamber. Temperature range is -25°C to 50°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Freeze-thaw chambers are excellent for customers requiring ramp/soak programming. They exceed ICH/FDA guidelines concerning uniformity requirements. Approximate ramping ability is as follows 1°C per 2 minutes from -23°C to +23°C and back, unloaded. Note: Some defrost tolerance may be required for extended operation (more than 48 hours) below 4°C.