Learn More
Felix Storch Inc ACCUCOLD The FS24L is a compact all-freezer designed to reach -20 C, the ideal temperature for long-term storage of select sensitive materials. The 1.4 cu.ft. manual defrost interior offers static cooling for stable results
SDP
Catalog No. 501959444
Encompass_Preferred
ACCUCOLD by SUMMIT Appliance offers a quality line of general purpose all-freezers designed for reliable cold storage in medical, scientific, and laboratory facilities. The FS24L is a compact all-freezer designed to reach -20 C, the ideal temperature for long-term storage of select sensitive materials. The 1.4 cu.ft. manual defrost interior offers static cooling for stable results. A removable wire shelf allows easy storage options, with a seamless interior liner to simplify cleanup. The dial thermostat is located inside. This unit has a flat door liner as recommended by the CDC: https://www.accucold.com/product/FS24L
Catalog No. 50-195-9444 Supplier Felix Storch Inc Supplier No. FS24L
ACCUCOLD by SUMMIT Appliance offers a quality line of general purpose all-freezers designed for reliable cold storage in medical, scientific, and laboratory facilities. The FS24L is a compact all-freezer designed to reach -20 C, the ideal temperature for long-term storage of select sensitive materials. The 1.4 cu.ft. manual defrost interior offers static cooling for stable results. A removable wire shelf allows easy storage options, with a seamless interior liner to simplify cleanup. The dial thermostat is located inside. This unit has a flat door liner as recommended by the CDC: https://www.accucold.com/product/FS24L