Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Freezer Rack 13 box capacity, 5 1/2 x 5 5/8 x 28 1/2 inches
Catalog No. NC1414844
Encompass_Preferred
13 Box Capacity; Dimensions (DxWxH): 5 1/2 x 5 5/8 x 28 1/2 inches (140 x 143 x 724mm); Compartment Dimensions: 5 1/2 x 5 1/2 x 2 3/16 inches (140 x 140 x 55mm)
Catalog No. NC1414844 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. CF132
13 Box Capacity; Dimensions (DxWxH): 5 1/2 x 5 5/8 x 28 1/2 inches (140 x 143 x 724mm); Compartment Dimensions: 5 1/2 x 5 1/2 x 2 3/16 inches (140 x 140 x 55mm)