Learn More
Boca Scientific Inc BOCARACK815 Freezer Storage System for 15ml Conical Tubes, Freezer Rack with Tube Storage Boxes, Capacity of 288 Tubes
Catalog No. NC1786405
Encompass
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in freezers, including low-temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and up-right freezers, including -80°C freezers. The Bocarack 815 contains 2 drawers with 4 polypropylene boxes in each drawer. Each box has a 6 x 6 configuration and a capacity of 36 15ml conical tubes. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.
Catalog No. NC1786405 Supplier Boca Scientific Inc Supplier No. BOCARACK815
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in freezers, including low-temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and up-right freezers, including -80°C freezers. The Bocarack 815 contains 2 drawers with 4 polypropylene boxes in each drawer. Each box has a 6 x 6 configuration and a capacity of 36 15ml conical tubes. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.