Learn More
Research Products International Corp Upright Freezer Rack for 2 Inch High Boxes, 20 Box Capacity, 4 x 5 Array, 22 1/8 x 5 1/2 x 11 1/2H (Inches)
SDP
Catalog No. 50476631
Encompass
Convenient storage system for 1.5ml - 2.0ml micro-tubes and 1.2ml - 2.0ml cryogenic vials. Stainless steel freezer racks are available for this upright freeze storage system. Racks include stainless steel handles and accommodate 2 inch high cardboard and metal storage boxes.
Catalog No. 50-476-631 Supplier Research Products International Corp Supplier No. 452
Convenient storage system for 1.5ml - 2.0ml micro-tubes and 1.2ml - 2.0ml cryogenic vials. Stainless steel freezer racks are available for this upright freeze storage system. Racks include stainless steel handles and accommodate 2 inch high cardboard and metal storage boxes.