Learn More
Research Products International Corp Chest Freezer Rack for 2 Inch High Boxes, 11 Box Capacity, 23 7/8H Inches
SDP
Catalog No. 50212137
Encompass
Convenient storage system for 1.5ml-2.0ml micro-tubes and 1.2ml-2.0ml cryogenic vials. Stainless steel freezer racks available for chest freezers. Racks include stainless steel handles and accommodate all 2" high cardboard and metal storage boxes. Separate components allow system design compatibility with space requirements.
Catalog No. 50-212-137 Supplier Research Products International Corp Supplier No. 1203
Convenient storage system for 1.5ml-2.0ml micro-tubes and 1.2ml-2.0ml cryogenic vials. Stainless steel freezer racks available for chest freezers. Racks include stainless steel handles and accommodate all 2" high cardboard and metal storage boxes. Separate components allow system design compatibility with space requirements.