Learn More
Research Products International Corp Upright Freezer Rack for Hinged-Top Plastic Boxes, 2 x 3 Array, 12 1/2 x 5 7/8 x 7 Inches
SDP
Catalog No. 50212548
Encompass
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.
Catalog No. 50-212-548 Supplier Research Products International Corp Supplier No. 247567
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.