Learn More
Research Products International Corp Upright Freezer Rack for Hinged-Top 100 Cell Plastic Boxes, 12 1/2 x 5 7/8 x 9 1/4 Inches
SDP
Catalog No. 50212546
Encompass
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.
Catalog No. 50-212-546 Supplier Research Products International Corp Supplier No. 247560
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.