Learn More
Electron Microscopy Sciences Electropolished Stainless Steel Freezer Rack 3x4, Holds 12 3" boxes
SDP
Catalog No. 501902282
Encompass_Preferred
High quality, ultra-durable freezer racks for long term sample storage. All edges have been ground down to remove the sharp surfaces that plague standard freezer racks. Electropolishing cleans up all weld points that, over time, may be the starting points for corrosion and leaves racks ultra-shiny and free from any surface contaminants. Superior construction and design have created a more sturdy rack than what is currently available on the market. Racks are designed and finished to last longer in the demanding freezer environment where samples are housed.
Capacity: Holds 12 3" boxes
Catalog No. 50-190-2282 Supplier Electron Microscopy Sciences Supplier No. 6161530
High quality, ultra-durable freezer racks for long term sample storage. All edges have been ground down to remove the sharp surfaces that plague standard freezer racks. Electropolishing cleans up all weld points that, over time, may be the starting points for corrosion and leaves racks ultra-shiny and free from any surface contaminants. Superior construction and design have created a more sturdy rack than what is currently available on the market. Racks are designed and finished to last longer in the demanding freezer environment where samples are housed.
Capacity: Holds 12 3" boxes