Learn More
Research Products International Corp UPRIGHT FREEZER RACK FOR HINGED TOP PLASTIC BOXES, 18 3/4 X 5 7/8 X 9 1/4 INCH (H)
SDP
Catalog No. 50212547
Encompass
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.
Catalog No. 50-212-547 Supplier Research Products International Corp Supplier No. 247561
The freezer rack for hinged boxes can hold 100 cell hinged-top plastic boxes. This rack provides a a durable alternative to the styrofoam or card racks. Saves freezer space and helps organize boxes.