Learn More
Electron Microscopy Sciences Arctic Squares Freezer Rack
SDP
Catalog No. 501929503
Encompass_Preferred
This freezer rack is constructed of corrosion-resistant stainless steel and features four drawers to hold 16 boxes. It is ideal for storing boxes up to 133 x 133 x 53 mm in size. The drawers pivot down when fully extended, allowing for easy box access. Polycarbonate drawer handles provide a user-friendly grip. Includes a slot and label for drawer/sample identification. Freezer rack includes a pull-out wire handle for easy removal off the freezer shelf. Fits most standard upright freezers.
Catalog No. 50-192-9503 Supplier Electron Microscopy Sciences Supplier No. 61612
This freezer rack is constructed of corrosion-resistant stainless steel and features four drawers to hold 16 boxes. It is ideal for storing boxes up to 133 x 133 x 53 mm in size. The drawers pivot down when fully extended, allowing for easy box access. Polycarbonate drawer handles provide a user-friendly grip. Includes a slot and label for drawer/sample identification. Freezer rack includes a pull-out wire handle for easy removal off the freezer shelf. Fits most standard upright freezers.