Learn More
Thomas Scientific Vertical Freezer Rack for 3" box, 5 cap
SDP
Catalog No. 501459179
Encompass_Preferred
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes
Catalog No. 50-145-9179 Supplier Thomas Scientific Supplier No. 1201Y64
Add to Cart
Add to Cart
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes