Learn More
Accuflow Systems Horizontal freezer rack 242
Catalog No. 501537791
Encompass_Preferred
Stainless Steel horizontal rack to hold 8 standard 2'' freezer boxes. Overall dimensions 11.25 X 8.75 X 5.50 (286 X 223 X 140MM)
Catalog No. 50-153-7791 Supplier Accuflow Systems Supplier No. HT82
Add to Cart
Add to Cart
Stainless Steel horizontal rack to hold 8 standard 2'' freezer boxes. Overall dimensions 11.25 X 8.75 X 5.50 (286 X 223 X 140MM)