Learn More
Labrepco Inc FREEZER RACK ONLY-FOR 2" BOXES
SDP
Catalog No. NC1539848
Encompass_Preferred
Value Series Upright Freezer Rack for Standard 2" Boxes- Rack Only (4 Boxes Deep x 5 Boxes High)Dimensions(LxHxW): 22.3" × 11.03" × 5.5"Capacity: 20 Boxes Per RackShipping Weight (lb): 8.5Includes: Rack Only
Catalog No. NC1539848 Supplier Labrepco Inc Supplier No. CUF452
Value Series Upright Freezer Rack for Standard 2" Boxes- Rack Only (4 Boxes Deep x 5 Boxes High)Dimensions(LxHxW): 22.3" × 11.03" × 5.5"Capacity: 20 Boxes Per RackShipping Weight (lb): 8.5Includes: Rack Only