Learn More
Research Products International Corp Freezer Rack, Upright, for 2"" Box, 5x4 Array
SDP
Catalog No. 501977660
Encompass
Freezer Rack in designed to hold 50 cell hinged-top plastic box. Fit 5 1/2 x 3 18 x 2 1/8 inch boxes. This rack provides a durable alternative to Styrofoam or card racks. Racks save freezer space and help organize boxes.
Catalog No. 50-197-7660 Supplier Research Products International Corp Supplier No. 247592
Freezer Rack in designed to hold 50 cell hinged-top plastic box. Fit 5 1/2 x 3 18 x 2 1/8 inch boxes. This rack provides a durable alternative to Styrofoam or card racks. Racks save freezer space and help organize boxes.