Learn More
Labrepco Inc FREEZER RACK,VERT,F/3"CRYO BOX
SDP
Catalog No. NC1451791
Encompass_Preferred
Vertical racks for Chest Freezers and Liquid Nitrogen Tanks. Racks are made of corrosion resistant stainless steel and include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.Specifications:Dimensions (HxWxD): 25 7/16" × 5 5/8" × 5 1/2" or 646 × 143 × 140mmCapacity: 8 boxes per rack*Rack only; Boxes and cell dividers not included.
Catalog No. NC1451791 Supplier Labrepco Inc Supplier No. CF83
Vertical racks for Chest Freezers and Liquid Nitrogen Tanks. Racks are made of corrosion resistant stainless steel and include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.Specifications:Dimensions (HxWxD): 25 7/16" × 5 5/8" × 5 1/2" or 646 × 143 × 140mmCapacity: 8 boxes per rack*Rack only; Boxes and cell dividers not included.