Learn More
Labrepco Inc FREEZER RACK W/DRAWER STD 2IN
SDP
Catalog No. NC1676575
Encompass_Preferred
Upright Freezer Rack with Drawers for Standard 2" Boxes-Rack Only (5 Boxes Deep x 5 Boxes High)High quality Upright Freezer Drawer Racks are constructedof corrosion resistant stainless steel, including handles andidentification card holders. Upright Freezer Drawer racksoffer a more organized system for storage and retrieval ofsamples. Drawer racks facilitate both access to andstorage of samples without having to remove the whole rackfrom the freezer.
Catalog No. NC1676575 Supplier Labrepco Inc Supplier No. UFD552
Upright Freezer Rack with Drawers for Standard 2" Boxes-Rack Only (5 Boxes Deep x 5 Boxes High)High quality Upright Freezer Drawer Racks are constructedof corrosion resistant stainless steel, including handles andidentification card holders. Upright Freezer Drawer racksoffer a more organized system for storage and retrieval ofsamples. Drawer racks facilitate both access to andstorage of samples without having to remove the whole rackfrom the freezer.