Learn More
USA Scientific Inc 20-place (4 x 5) upright freezer sliding rack for 2
Catalog No. NC0609122
Encompass
20-place (4 long x 5 high) upright freezer sliding rack for 2" H boxes, 22" L x 5 1/2" W x 11 13/16" H, stainless steel
Catalog No. NC0609122 Supplier USA Scientific Inc Supplier No. 26324500
20-place (4 long x 5 high) upright freezer sliding rack for 2" H boxes, 22" L x 5 1/2" W x 11 13/16" H, stainless steel