Description
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
- Epoxy-coated open-wire shelves
- Full- and half-depth sizes
- Undercounter units use full-depth shelves only
Specifications
Specifications
| Type | Open Wire Shelves |
| Exterior Finish | Epoxy-coated Steel |
| For Use With (Equipment) | 51.1 cu. ft. Units |
| Warranty | Warranty varies by country, please contact us for details |
| Holds | General Freezer Items |