Learn More
Research Products International Corp Freezer Rack for 25 Place Slides, Stainless Steel, 6 x 6 Array, Dimensions (Inches) 4 1/8 x 21 x 9 1/4H
SDP
Catalog No. 50550269
Encompass
The slotted Microscope Slide Freezer Rack provides convenient storage for slide racks. Helps save time by keeping you organized in a cost efficient manner. The highest quality racks, constructed of corrosion resistant stainless steel, includes handles. Racks for 25 place slide boxes fit all 3 13/16 x 3 1/4 x 1 5/16 boxes.
Catalog No. 50-550-269 Supplier Research Products International Corp Supplier No. 247523
The slotted Microscope Slide Freezer Rack provides convenient storage for slide racks. Helps save time by keeping you organized in a cost efficient manner. The highest quality racks, constructed of corrosion resistant stainless steel, includes handles. Racks for 25 place slide boxes fit all 3 13/16 x 3 1/4 x 1 5/16 boxes.