Learn More
Darwin Chambers Company Benchtop Stability Freezer 9 cu. ft.
SDP
Catalog No. 501893867
Encompass_Preferred
The FS011 is a 11.6 cubic foot benchtop stability freezer. Temperature range is -25°C to 15°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. Unsurpassed temperature control through continuous refrigeration bypass system. Auto-cycle defrost with short temperature rise.
Catalog No. 50-189-3867 Supplier Darwin Chambers Company Supplier No. FS011
Add to Cart
Add to Cart
Request a Quote
The FS011 is a 11.6 cubic foot benchtop stability freezer. Temperature range is -25°C to 15°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. Unsurpassed temperature control through continuous refrigeration bypass system. Auto-cycle defrost with short temperature rise.