Learn More
Darwin Chambers Company Triple Door Stability Freezer 66 cu. ft.
SDP
Catalog No. 501893931
Encompass_Preferred
The FS055-SS is a 50 cubic feet double door stability freezer. Temperature range is -25°C to 15°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Unsurpassed temperature control through continuous refrigeration bypass system. Auto-cycle defrost with short temperature rise.
Catalog No. 50-189-3931 Supplier Darwin Chambers Company Supplier No. FS055SS
Add to Cart
Add to Cart
Request a Quote
The FS055-SS is a 50 cubic feet double door stability freezer. Temperature range is -25°C to 15°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Unsurpassed temperature control through continuous refrigeration bypass system. Auto-cycle defrost with short temperature rise.