Learn More
Darwin Chambers Company Triple Door Stability Freezer 66 cu. ft.
SDP
Supplier:  Darwin Chambers Company FS084SS
Encompass_Preferred
The FS084-SS is a 76.2 cubic feet triple door stability freezer. Temperature range is -25°C to 15°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Unsurpassed temperature control through continuous refrigeration bypass system. Auto-cycle defrost with short temperature rise.
Catalog No. 50-189-3869
Add to Cart
Request a Quote