Learn More
Boca Scientific Inc BOCARACK850 Freezer Storage System for 50ml Conical Tubes, Freezer Rack with Tube Storage Boxes, Capacity of 128 Tubes
Catalog No. NC0016466
Encompass
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and upright freezers including -80°C freezers. The Bocarack 850 contains 2 drawers with 4 polypropylene boxes in each drawer. Each box has a 4 x 4 configuration and a capacity of 16 50ml conical tubes. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.
Catalog No. NC0016466 Supplier Boca Scientific Inc Supplier No. BOCARACK850
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include sliding drawers containing tube storage boxes. They are compatible with refrigerators and upright freezers including -80°C freezers. The Bocarack 850 contains 2 drawers with 4 polypropylene boxes in each drawer. Each box has a 4 x 4 configuration and a capacity of 16 50ml conical tubes. Numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. The rack measures 24" deep x 6" wide x 10" high.