Learn More
Sigma Aldrich Fine Chemicals Biosciences freezer storage box 81-pla
Catalog No. 502900304
Encompass
Sturdy freezer boxes provide compact storage and keep microcentrifuge tubes organized on the bench in the freezer or during transport. 81-place 9 x 9 row boxes hold 0.5 1.5 and 2.0 mL tubes and cryo vials.Alphanumeric indexingWithstand -80C to 121C (-112 to 249f)Stackable with molded-in no slide featuresLift off lid has matte panels on two sides for convenient labeling
Catalog No. 50-290-0304 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7566285EA
Sturdy freezer boxes provide compact storage and keep microcentrifuge tubes organized on the bench in the freezer or during transport. 81-place 9 x 9 row boxes hold 0.5 1.5 and 2.0 mL tubes and cryo vials.Alphanumeric indexingWithstand -80C to 121C (-112 to 249f)Stackable with molded-in no slide featuresLift off lid has matte panels on two sides for convenient labeling