Learn More
Chemglass Life Sciences Freezer Storage Box, Cardboard, Aluminum Foil Covered, 81 Cell, Complete with Dividers
SDP
Catalog No. 501532095
Encompass_Preferred
Freezer Storage Box, Cardboard, Aluminum Foil Covered, 81 Cell, Complete with Dividers. Durable storage boxes made of aluminum covered cardboard to withstand effects of liquid nitrogen and condensation from freezer equipment. Boxes can withstand temperatures as low as -196C. Designed to be compatible with most standard upright rack systems.
Catalog No. 50-153-2095 Supplier Chemglass Life Sciences Supplier No. CLS1198A81
Freezer Storage Box, Cardboard, Aluminum Foil Covered, 81 Cell, Complete with Dividers. Durable storage boxes made of aluminum covered cardboard to withstand effects of liquid nitrogen and condensation from freezer equipment. Boxes can withstand temperatures as low as -196C. Designed to be compatible with most standard upright rack systems.